M35606
α-Methylcinnamic acid
99%
Synonym(s):
alpha-Methylcinnamic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH=C(CH3)COOH
CAS Number:
Molecular Weight:
162.19
Beilstein:
2042546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
mp
79-81 °C (lit.)
SMILES string
C\C(=C/c1ccccc1)C(O)=O
InChI
1S/C10H10O2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-7H,1H3,(H,11,12)/b8-7+
InChI key
XNCRUNXWPDJHGV-BQYQJAHWSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service