Skip to Content
Merck
All Photos(1)

Key Documents

L050050

Sigma-Aldrich

Cap B1

configured for (ÄKTA® and OligoPilot®)

Synonym(s):

Cap B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

suitability

conforms to structure (GC, Formulation)

compatibility

configured for (ÄKTA® and OligoPilot®)

Looking for similar products? Visit Product Comparison Guide

General description

Cap B1 solution, containing acetonitrile (ACN) and acetic anhydride, is mixed in situ with Cap A and Cap B2 solution during the capping (acetylation) reaction. Pyridine is applied as a mild base while NMI provides a powerful acylation catalyst.

Preparation Note

Acetonitrile / Acetic Anhydrid 6/4 (v/v)

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service