GF75041760
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
foil, 10mm disks, 0.125mm thickness, annealed
Synonym(s):
FE240270
About This Item
Recommended Products
form
annealed
foil (disks)
manufacturer/tradename
Goodfellow 750-417-60
size × thickness
10 mm × 0.125 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | BML00261 | BML00141 | BML00287 |
---|---|---|---|
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service