GF62112181
Stainless steel - AISI 304
foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, diameter 10 mm, temper hard
Synonym(s):
FE220271
About This Item
Recommended Products
form
foil
manufacturer/tradename
Goodfellow 621-121-81
diam.
10 mm
thickness
0.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service