GF40319962
Stainless Steel - AISI 304 alloy, FeCr18Ni10
1000mm tube, 0.5mm outside diameter, 0.125mm wall thickness, 0.25mm inside diameter, hard
Synonym(s):
FE227150
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 403-199-62
L
1000 mm
O.D. × I.D. × thickness
0.5 mm × 0.125 mm × 0.25 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service