Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW445P

Azlon Square Shoulder Unvented Wash Bottles

wide-neck, low-density polyethylene bottle, red polypropylene closure, capacity 1000 mL

Synonym(s):

WGW445P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle
red polypropylene closure
wide-neck

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW445P

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Precision jet tip for fine control

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service