DWKW224682
WHEATON® amber sample vial with PTFE faced rubber lined cap packed in partitioned tray
glass, tube capacity (4 mL), screw cap
Synonym(s):
Sample Vial, W224682
All Photos(1)
About This Item
Recommended Products
material
amber borosilicate glass tube
flat bottom
glass
solid black phenolic cap
sterility
non-sterile
feature
with cap
packaging
case of 144 ea
manufacturer/tradename
WHEATON® W224682
tube capacity
(4 mL)
tube diam.
(15 mm)
fitting
neck (External Thread)
General description
Amber Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.
- Amber vials are manufactured from Wheaton 320 glass that conforms to USP Type I requirements to protect light-sensitive samples.
- Screw caps are attached to vials.
- It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples.
- Vials and caps are autoclavable.
- 2 mL vials fit the M-T Vial File, 228778.
- 4 mL vials fit the M-T Vial File, 228780.
Features and Benefits
• Clear vials are converted from Type I borosilicate glass tubing• Amber vials are converted from amber borosilicate glass tubing that conforms to USP Type I requirements for light transmission to protect light sensitive products• Sample vials with caps attached to vial help maintain cleanliness• Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials• Shorty Vial® feature a low profile for greater stability and less dead volume• 14B rubber lined black phenolic screw cap for aqueous samples• PTFE / 14B rubber lined black phenolic screw cap for organic samples• Replacement caps can be purchased separately
Legal Information
Lab File is a registered trademark of Wheaton Industries, Inc.
Shorty Vial is a registered trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)