DWK74504-20
KIMBLE® scintillation vials with attached pulp backed foil lined PP cap
lips on vial transparent borosilicate glass bottle, vial capacity (20 mL), screw cap
All Photos(1)
About This Item
Recommended Products
material
lips on vial transparent borosilicate glass bottle
metal foil liner
white polypropylene cap
sterility
non-sterile
feature
class A
with cap
packaging
case of 500 ea 5x shrink-wrapped trays of 100 vials
manufacturer/tradename
Kimble® 74504-20
bottle diam. × H
(27 mm) (57 mm)
vial capacity
(20 mL)
fitting
22-400 neck
22-400
General description
KIMBLE® glass scintillation vials feature 20mL borosilicate glass vials with polypropylene caps that are securely attached. These vials are packaged in cellular trays that feature alphanumeric coordinates, enabling quick and effortless identification.
Features and Benefits
• Converted from Type I, 33 expansion borosilicate glass tubing.• Each tray contains 100 vials• Five trays per case, shrink-wrapped to preserve cleanliness• Caps attached
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)