DWK218052959
Duran® Protect, Laboratory Bottle
Laboratory bottle, plastic-coated glass bottle, neck Joints: threaded (GL45), capacity 150 mL, with screw cap to drain contents, WITH SCREW-CAP AND POURING RING (PP)
Synonym(s):
218052959
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
plastic-coated glass bottle
Joint
Joints: threaded neck (GL45)
feature
with screw cap to drain contents WITH SCREW-CAP AND POURING RING (PP)
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 218052959
capacity
150 mL
Looking for similar products? Visit Product Comparison Guide
General description
DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134 °C.
Application
Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Liquid handling reservoir, Boro 3.3, plastic coated
Features and Benefits
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, plastic coated Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic print; Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service