Skip to Content
Merck
All Photos(5)

Key Documents

D163708

Sigma-Aldrich

N,O-Dimethylhydroxylamine hydrochloride

98%

Synonym(s):

N,O-Dimethylhydroxylamine HCl, N-Methoxymethylamine hydrochloride, Methoxymethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ONHCH3 · HCl
CAS Number:
Molecular Weight:
97.54
Beilstein:
3650353
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

112-115 °C

SMILES string

Cl[H].CNOC

InChI

1S/C2H7NO.ClH/c1-3-4-2;/h3H,1-2H3;1H

InChI key

USZLCYNVCCDPLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the preparation of Weinreb amides recently used in the synthesis of 2-acyloxazoles from 2-oxazolemagnesium chloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Pippel et al.
The Journal of organic chemistry, 72(15), 5828-5831 (2007-06-26)
Treatment of oxazole or 5-aryl oxazoles with i-PrMgCl smoothly generates the corresponding 2-Grignard reagents, which react with Weinreb amides to provide exclusively 2-acyl oxazole products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service