CLS9944510
Pyrex® disposable culture tubes, rimless
capacity 4.0 mL
Synonym(s):
culture tube, test tubes, Corning® 99445
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
description
dispenser arrangement
sterility
non-sterile
feature
cap: no
packaging
pkg of 1000 ea
manufacturer/tradename
Corning 99445-10
O.D. × L
10 mm × 75 mm
capacity
4.0 mL
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Single-use culture tubes are convenience packed.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service