CAD9881
Dissolution cannulae
gauge 14, 316 stainless steel hub, L 7 1/2 in.
All Photos(1)
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
Recommended Products
material
316 stainless steel hub
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 9881
L
7 1/2 in.
gauge
14
Related Categories
General description
Ideal for drug dissolution testing. Drug dissolution testing is routinely used in the pharmaceutical industry to provide critical release information for quality control purposes, in new drug development, and to predict in vivo drug release profiles. These cannula feature 316 stainless steel hub and tubing for maximum resistance to chemical reaction.
Choose from one of the most recent versions:
Certificates of Analysis (COA)