CAD6056
3-way stopcock
female Luer lock to female Luer lock, male Luer lock, in. Y in. type
All Photos(1)
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 6056
Related Categories
General description
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
- Made from nickel plated brass or stainless steel
- Tested to 45 psi water pressure
- High pressure stopcocks are rated to 290 psi
Choose from one of the most recent versions:
Certificates of Analysis (COA)