All Photos(1)
About This Item
Linear Formula:
ClC6H4NHNH2 · HCl
CAS Number:
Molecular Weight:
179.05
Beilstein:
3596254
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
powder
mp
216 °C (dec.) (lit.)
SMILES string
Cl.NNc1ccc(Cl)cc1
InChI
1S/C6H7ClN2.ClH/c7-5-1-3-6(9-8)4-2-5;/h1-4,9H,8H2;1H
InChI key
YQVZREHUWCCHHX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J P Mahy et al.
European journal of biochemistry, 226(2), 445-457 (1994-12-01)
The reaction of p-chlorophenylhydrazine with prostaglandin H synthase (PGHS) Fe(III) under aerobic conditions leads to a partial destruction of the heme and to a new complex absorbing at 436 nm. This complex is also obtained by reaction of p-chlorophenyldiazene (pClPhN
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service