All Photos(1)
About This Item
Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.557 (lit.)
bp
147 °C/25 mmHg (lit.)
mp
21 °C (lit.)
density
1.298 g/mL at 25 °C (lit.)
SMILES string
Cc1cccc(c1Cl)[N+]([O-])=O
InChI
1S/C7H6ClNO2/c1-5-3-2-4-6(7(5)8)9(10)11/h2-4H,1H3
InChI key
XTSGZXRUCAWXKY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service