Skip to Content
Merck
All Photos(2)

Key Documents

C105805

Sigma-Aldrich

Cyclohexanemethanol

ReagentPlus®, ≥99%

Synonym(s):

(Hydroxymethyl)cyclohexane, Cyclohexyl methanol, Hexahydrobenzyl alcohol, NSC 5288

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2OH
CAS Number:
Molecular Weight:
114.19
Beilstein:
773712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

≥99%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

181 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCCC1

InChI

1S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8H,1-6H2

InChI key

VSSAZBXXNIABDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cyclohexanemethanol (Cyclohexyl methanol) can be used as a starting material for the synthesis of cyclohexanecarboxaldehyde, cyclohexanecarboxylic acid, cyclohexanone, and 1,4-cyclohexadione by photocatalytic oxidation (PCO) using titanium dioxide nanoparticles as a catalyst.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoparticles TiO2-photocatalyzed oxidation of selected cyclohexyl alcohols.
Mohamed OS, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 200(2-3), 209-215 (2008)
Yasunori Okada et al.
Organic letters, 9(8), 1573-1576 (2007-03-16)
[reaction: see text] Ethyl 1-thio-2,3,4,6-tetrakis-O-triisopropylsilyl-beta-d-glucopyranoside, ethyl 6-O-benzyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside, and ethyl 6-O-pivaloyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside induced highly beta-selective O-glucosidations. Among them, the 6-O-pivaloylated substrate provided the best selectivity up to alpha/beta = 3:97 with cyclohexylmethanol, and the substrate was used for glucosidations with secondary and
Rabea Schlüter et al.
Applied microbiology and biotechnology, 103(10), 4137-4151 (2019-04-04)
The cycloalkanes, comprising up to 45% of the hydrocarbon fraction, occur in crude oil or refined oil products (e.g., gasoline) mainly as alkylated cyclohexane derivatives and have been increasingly found in environmental samples of soil and water. Furthermore, short-chain alkylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service