Key Documents
BR759105
BRAND® macro-cuvette
PMMA
Synonym(s):
Cuvettes
About This Item
UNSPSC Code:
41121813
NACRES:
SB.32
material
PMMA
feature
minimum filling volume 2.5 mL
manufacturer/tradename
BRAND 759105
General description
  • BRAND® Macro cuvette is manufactured under controlled room conditions.
  • Ideal for the determinations in water analysis, chemistry, and life science applications.
  • Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Eliminates the risk of cross-contamination.
  • Grouped by mold cavity number.
  • An economical alternative to Quartz cuvettes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)