Key Documents
BR749321
BRAND® micro haematocrit capillary
non-hep.
Synonym(s):
Haematocrit capillaries
About This Item
UNSPSC Code:
41121500
NACRES:
NB.22
material
blue
feature
DIN ISO 12772 and BS 4316-68, To Contain
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 749321
General description
Compliant with DIN ISO 12772 and BS 4316-68 standards, these tubes feature a color code for easy identification and use in hematocrit centrifuges. The heparinized tubes are treated with sodium heparinate on the internal surface and are marked with a red color code, making them suitable for capillary blood samples. In contrast, the non-heparinized tubes are designated with a blue color code, specifically designed for use with heparinized venous blood.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Guang Yang et al.
Journal of cellular and molecular medicine, 19(10), 2413-2422 (2015-07-16)
Previous work from both our lab and others have indicated that exposure to 50 Hz magnetic fields (ELF-MF) was able to modify ion channel functions. However, very few studies have investigated the effects of MF on γ-aminobutyric acid (GABA) type