Key Documents
BR701968
BRAND® repair set for TRANSFERPETTOR
for 100/200 μL
Synonym(s):
Accessories, Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor
About This Item
UNSPSC Code:
41100000
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
BRAND 701968
Related Categories
General description
Includes: 1 allen key, 1 piston rod with fitted PTFE Transferpettor-seal (above size 20 μL), 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 Transferpettor-seals, PTFE 1 mounting block (above size 20 μL).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)