BMSC33031215
Benchmark 12 x 15ml fixed angle rotor for ”Sprint” 6H Plus
Benchmark 12 x 15ml fixed angle rotor for Sprint 6H Plus
Synonym(s):
centrifuge rotor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
to hold 12 x 15 ml
manufacturer/tradename
Benchmark Scientific C3303-1215
parameter
4000 rpm speed
compatibility
Sprint 6H Plus
General description
Optional 12 x 15ml fixed angle rotor for Sprint 6H Plus
Application
Clinical centrifuge, centrifuge, research centrifuge, LC8, 15ml, 10ml, vacutainer, blood tube
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service