Key Documents
BAF379200000
SP Bel-Art® Plastic Forceps
polypropylene, L 5 in. (127 mm)
Synonym(s):
plastic forcep, reusable forceps
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
polypropylene
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art F37920-0000
L
5 in. (127 mm)
General description
These polypropylene plastic forceps have serrated tips to ensure a firm grasp.
Features and Benefits
  • Strong and durable
  • Steam autoclavable up to 121ºC (250ºF)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)