Key Documents
BAF182060000
SP Bel-Art® Double Burette Clamp
for 0.5 in. Support Rod
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
feature
to hold Double
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18206-0000
General description
Non-corroding clamps hold burettes firmly in place.
Features and Benefits
  • Soft rubber pads hold burette without scratching
  • Clamp fits standard 12.5mm (½") support rod and includes an adapter to fit to narrower 9.5mm (⅜") rod (support rod not included)
  • Made of chemical resistant polypropylene; autoclavable at 121°C (250°F)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)