BAF175700000
Sample vials with captive closure
capacity 0.13 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
colorless
colorless tube
flat bottom
low-density polyethylene cap
low-density polyethylene tube
sterility
non-sterile
feature
graduations: no
neck thread push fit
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F17570-0000
O.D. × H × I.D.
0.28 in. × 0.54 in. × 7 mm
capacity
0.13 mL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
These cylindrical vials with captive cap are made from lab-grade low density polyethylene (LDPE) to ensure the integrety of samples for testing. The vials are perfect for use in cleanrooms and have excellent chemical resistance.
- Snug fitting snap-on lid opens and closes with a flip of your thumb for single-handed operation
- Lab Grade LDPE won′t contaminate samples
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service