Key Documents
BAF131900100
Clavies® bag clips
autoclave safe
Synonym(s):
bag seal
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F13190-0100
General description
Open and close the clip with one hand. After autoclaving, clip holds the processed bag closed for final disposal. The closed internal dimension of the clip is 7/16 in., suitable for autoclave bags from 12 x 24 in. to 19 x 23 in. Do not use with larger bags.
  • Holds filled autoclave bags closed during cycle
  • Clip permits bag to safely vent internal pressure during autoclaving
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)