Skip to Content
Merck
All Photos(1)

Key Documents

AFM-71856-02

Sigma-Aldrich

Muscovite Mica for AFM

(V-1 Quality), diam. 12 mm, pack of 10

Synonym(s):

Muscovite Mica for AFM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.23

form

solid

packaging

pack of 10

diam.

12 mm

thickness

0.15-0.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

Mica, 12mm diameter, thickness 0.15-0.2 mm, V-1 grade, Clear - Hard, of uniform color, nearly flat, free of all stains, foreign inclusion, cracks, and other similar defects. Perfect for AFM substrate

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service