Key Documents
ACE6893015
Ace receiving flask for rotary evaporators, graduated, round bottom
size 500 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
round bottom
single-neck
Joint
joint 35/20
packaging
pkg of 1 ea
manufacturer/tradename
Ace Glass 6893-15
size
500 mL
General description
Graduated, round bottom receiving flask for use with all rotary evaporators. 35/20 spherical outer joint will also mate to 35/25 ball joints. 250 mL and 500 mL size graduated in 10 mL increments, 1000 mL and 2000 mL in 100 mL increments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)