Skip to Content
Merck
All Photos(1)

Key Documents

A52153

Sigma-Aldrich

6-Amino-1,3-dimethyluracil

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

295 °C (dec.) (lit.)

SMILES string

CN1C(N)=CC(=O)N(C)C1=O

InChI

1S/C6H9N3O2/c1-8-4(7)3-5(10)9(2)6(8)11/h3H,7H2,1-2H3

InChI key

VFGRNTYELNYSKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wafaa S Hamama et al.
Journal of advanced research, 4(2), 115-121 (2013-03-01)
The reaction of 6-amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione (1) as a binucleophile with primary aromatic or heterocyclic amines and formaldehyde or aromatic (heterocyclic) aldehydes in a molar ratio (1:1:2) gave the pyrimido[4,5-d]pyrimidin-2,4-dione ring systems 2-5. Treatment of 1 with diamines and formalin in molar
Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service