Skip to Content
Merck
All Photos(1)

Key Documents

936146

Sigma-Aldrich

RUMeIPr

≥95%

Synonym(s):

2-(2,6-Diisopropylphenyl)-5-(dimethylamino)-2H-imidazo[1,5-a]pyridin-4-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28ClN3
CAS Number:
Molecular Weight:
357.92
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

SMILES string

CC(C1=CC=CC(C(C)C)=C1N2C=[N+]3C(C=CC=C3N(C)C)=C2)C.[Cl-]

Application

RUMeIPr is a NHC ligand developed by the Szostak Group. This NHC analogue of the priviledged DalPhos scaffold has been used in the following reactions: Au-catalyzed C-C coupling, Au-catalyzed C-N coupling, and Au-catalyzed hydroamination of internal alkynes.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service