928666
SItHept HCl
≥95.0%
Synonym(s):
1,3-Bis(3-ethylpentan-3-yl)-4,5-dihydro-1H-imidazol-3-ium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Related Categories
Application
SItHept HCl is an N-alkyl NHC ligand with demonstrated uses in the following reactions. Au-catalyzed hydration, Cu-catalyzed hydroboration, Ag-catalyzed hydroboration, Cu-catalyzed C–O coupling and Pd-catalyzed C–C and C–N couplings. This ligand features significantly higher steric volume compared with ItBu (659983) while maintaining the σ-donation and σ-acceptance inherent to N-alkyl NHC ligands.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service