922501
TXO-TPA
≥99% (HPLC)
Synonym(s):
2-[4-(Diphenylamino) phenyl]-10, 10-dioxide-9H-thioxanthen-9-one
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C31H21NO3S
CAS Number:
Molecular Weight:
487.57
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
≥99% (HPLC)
solubility:
toluene: soluble
Recommended Products
Quality Level
Assay
≥99% (HPLC)
energy gap
0.052 eV, delta EST
loss
0.5% TGA, > 250 °C (weight loss)
solubility
toluene: soluble
fluorescence
λem 625 nm (film)
Orbital energy
HOMO -5.4 eV
LUMO -3.5 eV
λ
in toluene
UV absorption
λ: 299 nm±5 nm Amax
λ: 415 nm±5 nm Amax
Related Categories
Application
Sublimed high purity material used as a dopant for orange-red TADF OLED.
EQE of 9.75 %.
EQE of 9.75 %.
TXO-TPA can be employed as a hole transport layer in organic light-emitting diodes (OLEDs) and organic photovoltaic (OPV) devices. This material exhibits good charge transport properties, contributing to the overall performance and efficiency.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Manish Kumar et al.
ACS omega, 5(5), 2196-2204 (2020-02-18)
Charge balance, concentration quenching, and exciton confinement are the most important factors for realizing the use of thermally activated delayed fluorescence (TADF) emitters for organic light-emitting diodes. Red-orange organic light-emitting diodes of a TADF emitter 2-[4 (diphenylamino)phenyl]-10,10-dioxide-9H-thioxanthen-9-one (TXO-TPA) have been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service