Skip to Content
Merck
All Photos(1)

Key Documents

921734

Sigma-Aldrich

L-Ascorbic Acid-1-13C,4,5,6,6-d4

≥97 atom % D, ≥99 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5D4H4O6
Molecular Weight:
181.14
UNSPSC Code:
12352106

isotopic purity

≥99 atom % 13C
≥97 atom % D

Assay

≥97% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service