910112
Caffeine-(1-methyl-13C)
endotoxin tested, ≥99 atom % 13C, ≥99% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Assay
≥99% (CP)
form
solid
technique(s)
mass spectrometry (MS): suitable
suitability
endotoxin tested
mass shift
M+1
Application
For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the enduser.Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service