Skip to Content
Merck
All Photos(1)

Key Documents

900595

Sigma-Aldrich

L-Arginine-5-13C,4,4,5,5-d4 hydrochloride

≥99 atom % 13C, ≥97 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5D4H10N4O2 · HCl
Molecular Weight:
215.68
UNSPSC Code:
12352101

isotopic purity

≥99 atom % 13C
≥97 atom % D

Assay

≥98% (CP)

form

solid

mass shift

M+5

SMILES string

OC([C@@H](N)CC([2H])([2H])[13C]([2H])([2H])NC(N)=N)=O.[H]Cl

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service