900538
Ir(p-F-ppy)3
Synonym(s):
(OC-6-22)-Tris[5-fluoro-2-(2-pyridinyl-κN)phenyl-κC]iridium
About This Item
Recommended Products
form
powder or chunks
Quality Level
reaction suitability
core: iridium
reagent type: catalyst
reaction type: Photocatalysis
photocatalyst activation
465 nm
SMILES string
[Ir]741([NH]8=CC=CC=C8c9c7cc(cc9)F)([NH]5=CC=CC=C5c6c4cc(cc6)F)[NH]2=CC=CC=C2c3c1cc(cc3)F
InChI key
ZQIVPBRTCYGMPT-UHFFFAOYSA-N
Related Categories
Application
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service