Skip to Content
Merck
All Photos(1)

Key Documents

807591

Sigma-Aldrich

Silver

Nanowires, diam. × L 60 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141740
NACRES:
NA.23

description

Flash point >230 °F

Quality Level

form

suspension

concentration

5 mg/mL (in water)

diam. × L

60 nm (±10 nm) × 40 μm (±5 μm)

color

gray

density

1.0035 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to unique physical and chemical properties the nanowires of silver find wide range of applications extending from catalysis to photovoltaics, touch screen display, biosensing etc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service