799254
Sodium Glycolate-1-13C
99 atom % 13C, 98% (CP)
Synonym(s):
Glycolic-1-13C acid sodium
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
98% (CP)
form
powder
mass shift
M+1
storage temp.
2-8°C
SMILES string
[O-][13C](CO)=O.[Na+]
InChI
1S/C2H4O3.Na/c3-1-2(4)5;/h3H,1H2,(H,4,5);/q;+1/p-1/i2+1;
InChI key
VILMUCRZVVVJCA-CGOMOMTCSA-M
1 of 4
This Item | SU860079 | SU860092 | 507415 |
---|---|---|---|
PTFE/silicone septum, clear cap | blue polypropylene cap, red PTFE/silicone/red PTFE septum | blue polypropylene cap (with 6 mm center hole), red PTFE/white silicone septum | red polypropylene cap, tan PTFE/silicone septum |
55 (Durometer: shore A) | 45 (Durometer: shore A) | 55 (Durometer: shore A) | - |
pkg of 1000 ea | pkg of 100 ea | pkg of 100 ea | pkg of 100 ea |
9 mm × 1.0 mm | 9 mm × 1.0 mm | 9 mm × 1.0 mm | - |
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service