Skip to Content
Merck
All Photos(1)

Key Documents

792578

Sigma-Aldrich

Methyl 1-Boc-azetidine-3-carboxylate

95%

Synonym(s):

1-tert-Butyl 3-methyl azetidine-1,3-dicarboxylate, 1-Boc-azetidine-3-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.452

density

1.072 g/mL at 25 °C

functional group

ester

SMILES string

CC(C)(C)OC(N(C1)CC1C(OC)=O)=O

InChI

1S/C10H17NO4/c1-10(2,3)15-9(13)11-5-7(6-11)8(12)14-4/h7H,5-6H2,1-4H3

InChI key

SECFRXGVLMVUPD-UHFFFAOYSA-N

Application

Building block used as a precursor to the synthesis of azaspiro[3.4]octanes developed in the Carreira group. This substrate can be readily functionalized via enolization at the 3-position in the presence of LDA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service