Skip to Content
Merck
All Photos(2)

Key Documents

778923

Sigma-Aldrich

N-Boc-4-pentyne-1-amine

≥95% (GC)

Synonym(s):

4-Pentynylcarbamic acid tert-butyl ester, N-Boc-5-amino-1-pentyne, tert-Butyl (4-pentynyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO2
CAS Number:
Molecular Weight:
183.25
Beilstein:
6918435
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (GC)

form

liquid

reaction suitability

reaction type: click chemistry

SMILES string

CC(C)(C)OC(=O)NCCCC#C

InChI

1S/C10H17NO2/c1-5-6-7-8-11-9(12)13-10(2,3)4/h1H,6-8H2,2-4H3,(H,11,12)

InChI key

JTPJJKZSKWNWKK-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service