Skip to Content
Merck
All Photos(1)

Key Documents

766127

Sigma-Aldrich

2,5-Dilfuoro-4-methoxyphenylzinc bromide solution

0.6 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrF2OZn
Molecular Weight:
288.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

core: zinc
reagent type: catalyst
reaction type: Negishi Coupling

reagent type: derivatization reagent
reaction type: Negishi Coupling

concentration

0.6 M in THF

density

1.029 g/mL at 25 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

COc1cc(F)c([Zn]Br)cc1F

InChI

1S/C7H5F2O.BrH.Zn/c1-10-7-4-5(8)2-3-6(7)9;;/h3-4H,1H3;1H;/q;;+1/p-1

InChI key

WEUAQJYCDOJDPB-UHFFFAOYSA-M

Application

Nucleophlic reagent applicable for substitution and addition reactions. Also could be used in cross coupling reactions (Negishi couplings).

Organozinc Reagents

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service