760501
Diethyl-d10 carbonate
98 atom % D, 97% (CP)
Synonym(s):
Deuterium labeled diethyl carbonate, Diethyl carbonate-d10
About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
97% (CP)
form
liquid
bp
126-128 °C (lit.)
mp
-43 °C (lit.)
density
1.056 g/mL
mass shift
M+10
SMILES string
[2H]C([2H])([2H])C([2H])([2H])OC(=O)OC([2H])([2H])C([2H])([2H])[2H]
InChI
1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
InChI key
OIFBSDVPJOWBCH-MWUKXHIBSA-N
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
77.0 °F - closed cup
Flash Point(C)
25 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service