Skip to Content
Merck
All Photos(2)

Key Documents

756733

Sigma-Aldrich

Di-μ-chlorobis[2′-(amino-N)[1,1′-biphenyl]-2-yl-C]dipalladium(II)

Synonym(s):

Bis[2′-(amino-κN)[1,1′-biphenyl]-2-yl-κ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20Cl2N2Pd2
CAS Number:
Molecular Weight:
620.18
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

impurities

≤15% Tetrahydrofuran

mp

191-207 °C

SMILES string

Nc1ccccc1-c2ccccc2[Pd]Cl.Nc3ccccc3-c4ccccc4[Pd]Cl

InChI

1S/2C12H10N.2ClH.2Pd/c2*13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;;;/h2*1-6,8-9H,13H2;2*1H;;/q;;;;2*+1/p-2

InChI key

HUPALGDXJMOSBC-UHFFFAOYSA-L

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service