755028
5-Bromo-3-fluoropyridine-2-carbonitrile
97%
Synonym(s):
5-Bromo-3-fluoropicolinonitrile, 5-Bromo-2-cyano-3-fluoropyridine, 5-Bromo-3-fluoro-2-pyridinecarbonitrile
About This Item
Recommended Products
Assay
97%
form
solid
mp
95-100 °C
functional group
bromo
fluoro
nitrile
storage temp.
−20°C
SMILES string
Fc1cc(Br)cnc1C#N
InChI
1S/C6H2BrFN2/c7-4-1-5(8)6(2-9)10-3-4/h1,3H
InChI key
HMURQOFNWZWERT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service