Skip to Content
Merck
All Photos(1)

Key Documents

739995

Sigma-Aldrich

3-Methoxyphenylboronic acid pinacol ester

Synonym(s):

2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19BO3
CAS Number:
Molecular Weight:
234.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.06

form

liquid

Quality Level

refractive index

n20/D 1.502

density

1.028 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COc1cccc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C13H19BO3/c1-12(2)13(3,4)17-14(16-12)10-7-6-8-11(9-10)15-5/h6-9H,1-5H3

InChI key

DMKREZLZDOSDQU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service