Skip to Content
Merck
All Photos(3)

Key Documents

73178

Sigma-Aldrich

7-Bromo-3-hydroxy-2-naphthoic acid

≥98.0% (HPLC), 98.0-102.0 wt. % (AT)

Synonym(s):

7-Bromo-3-hydroxy-2-naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7BrO3
CAS Number:
Molecular Weight:
267.08
Beilstein:
2372925
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)
98.0-102.0% (wt., AT)

form

powder

functional group

bromo

SMILES string

OC(=O)c1cc2cc(Br)ccc2cc1O

InChI

1S/C11H7BrO3/c12-8-2-1-6-5-10(13)9(11(14)15)4-7(6)3-8/h1-5,13H,(H,14,15)

InChI key

XZWXQSGFZHRDNB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service