Recommended Products
isotopic purity
99 atom % 13C
Assay
98% (CP)
form
liquid
bp
99 °C (lit.)
mp
-73 °C (lit.)
density
0.896 g/mL at 25 °C
mass shift
M+1
SMILES string
CCO[13C](=O)CC
InChI
1S/C5H10O2/c1-3-5(6)7-4-2/h3-4H2,1-2H3/i5+1
InChI key
FKRCODPIKNYEAC-HOSYLAQJSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service