Skip to Content
Merck
All Photos(1)

Key Documents

719277

Sigma-Aldrich

Trichloroethylene-13C2

99 atom % 13C, 97% (CP)

Synonym(s):

TCE-13C2, Trichloroethene-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClH13C=13CCl2
CAS Number:
Molecular Weight:
133.37
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

bp

86.7 °C (lit.)

density

1.485 g/mL at 25 °C

mass shift

M+2

SMILES string

Cl\[13CH]=[13C](\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H/i1+1,2+1

InChI key

XSTXAVWGXDQKEL-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocio Aranda-Rodriguez et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 992, 76-85 (2015-05-13)
Humans are continuously exposed to volatile organic compounds (VOCs) as these chemicals are ubiquitously present in most indoor and outdoor environments. In order to assess recent exposure to VOCs for population-based studies, VOCs are measured in the blood of participants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service