All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H5BF3K
CAS Number:
Molecular Weight:
135.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
mp
80-84 °C
SMILES string
[K+].CC[B-](F)(F)F
InChI
1S/C2H5BF3.K/c1-2-3(4,5)6;/h2H2,1H3;/q-1;+1
InChI key
GIIPADVFWNGSKY-UHFFFAOYSA-N
Application
Potassium ethyltrifluoroborate can be used as a substrate in Suzuki–Miyaura coupling reaction with alkenyl bromides to produce alkene derivatives.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gary A Molander et al.
Tetrahedron, 63(3), 768-775 (2008-01-16)
The Suzuki-Miyaura-type cross-coupling reaction of potassium alkyltrifluoroborates with various alkenyl bromides in the presence of 10 mol % of PdCl(2)(dppf)·CH(2)Cl(2) and 3.0 equiv of Cs(2)CO(3) in aqueous toluene at 80 °C provided the desired compounds in 49-95% yields. A variety
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service