Skip to Content
Merck
All Photos(1)

Key Documents

707538

Sigma-Aldrich

(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethylenediamine

97%

Synonym(s):

(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18F6N2
CAS Number:
Molecular Weight:
376.34
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

113-115 °C (lit.)
114-118 °C (lit.)

SMILES string

CN[C@H]([C@@H](NC)c1cccc(c1)C(F)(F)F)c2cccc(c2)C(F)(F)F

InChI

1S/C18H18F6N2/c1-25-15(11-5-3-7-13(9-11)17(19,20)21)16(26-2)12-6-4-8-14(10-12)18(22,23)24/h3-10,15-16,25-26H,1-2H3/t15-,16-/m0/s1

InChI key

SBGOGHODWUZHIY-HOTGVXAUSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service