Skip to Content
Merck
All Photos(1)

Key Documents

705373

Sigma-Aldrich

1,7-Dimethylxanthine-(dimethyl-d6)

≥98 atom % D, ≥97% (CP)

Synonym(s):

1,7-Dimethyl-1H-purine-2,6-dione-(dimethyl-d6), 2,6-Dihydroxy-1,7-dimethylpurine-(dimethyl-d6), Paraxanthine-(dimethyl-d6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D6H2N4O2
CAS Number:
Molecular Weight:
186.20
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

294-296 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1C(=O)Nc2ncn(c2C1=O)C([2H])([2H])[2H]

InChI

1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)11(2)7(13)9-5/h3H,1-2H3,(H,9,13)/i1D3,2D3

InChI key

QUNWUDVFRNGTCO-WFGJKAKNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service