Skip to Content
Merck
All Photos(2)

Key Documents

700878

Sigma-Aldrich

4-Phenoxybenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9ClO3S
CAS Number:
Molecular Weight:
268.72
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

41-44 °C

functional group

phenoxy

SMILES string

ClS(=O)(=O)c1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9ClO3S/c13-17(14,15)12-8-6-11(7-9-12)16-10-4-2-1-3-5-10/h1-9H

InChI key

QIZPONOMFWAPRR-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
272248700894283509
form

powder

form

-

form

solid

form

liquid

mp

41-44 °C

mp

66-70 °C (lit.)

mp

60-61 °C

mp

-

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service